Learn More
Ready America Emergency Water Packet
Individual ready to drink purified water pouches with 5 year shelf life
Supplier: Ready America 73021
Description
- 5 year shelf life from manufacture date
- Produced with unique seven-layer carton
- Includes bendable drinking straws
- No risk of packing migration or unsavory taste
- Unique design allows for more water storage per inc
- Packaging is designed to conveniently stack multiple pouches in a compact fashion
- Purified by reverse osmosis to meet U.S. Pharmacopoeia standards and sealed in leak-proof pouches to prevent bacteria growth
Specifications
| Emergency Water | |
| 4 x 6 x 1 in. | |
| 0.27 lb. |