Learn More
Electron Microscopy Sciences EMF 10 Replacement Odor Element
SDP
Catalog No. 501926574
Encompass_Preferred
Replacement part for the EMF10 oil mist filter. The EMF10 is used with the Edwards RV3, RV5, and RV8 vacuum pumps.
Catalog No. 50-192-6574 Supplier Electron Microscopy Sciences Supplier No. 91005ROM
Replacement part for the EMF10 oil mist filter. The EMF10 is used with the Edwards RV3, RV5, and RV8 vacuum pumps.