Learn More
Electron Microscopy Sciences Edwards EMF 3 Replacement Mist Element
SDP
Catalog No. 501902456
Encompass_Preferred
Replacement part for the EMF3 oil mist filter. The EMF3 is used with the Edwards E2M1.5 vacuum pump.
Catalog No. 50-190-2456 Supplier Electron Microscopy Sciences Supplier No. 91004RMO
Replacement part for the EMF3 oil mist filter. The EMF3 is used with the Edwards E2M1.5 vacuum pump.