Learn More
Electron Microscopy Sciences Edwards EMF 3 Replacement Odor Element
SDP
Catalog No. 501926573
Encompass_Preferred
Replacement part for the EMF3 oil mist filter. The EMF3 is used with the Edwards E2M1.5 vacuum pump.
Catalog No. 50-192-6573 Supplier Electron Microscopy Sciences Supplier No. 91004ROM
Replacement part for the EMF3 oil mist filter. The EMF3 is used with the Edwards E2M1.5 vacuum pump.