Learn More
Medchemexpress LLC HY-131067 5mg Medchemexpress, EMI56 CAS:2414374-41-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1310675MG
Encompass_Preferred
Medchemexpress, HY-131067 5mg EMI56 CAS:2414374-41-5 EMI56, the derivative of EMI1, displays greater potency toward mutant EGFR than EMI1. EMI56 inhibits EGFR triple mutants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1263