Learn More
Aviva Systems Biology Corp EMID2 Antibody - C-terminal region (ARP52562_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52562_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EMID2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human EMID2. Alternate names: COL26A1, EMI6, EMU2, MGC129848, hEmu2, SH2B, EMID2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60772