Learn More
Cayman Chemical EmIdeltIdeacetate 50mg
SDP
Catalog No. 502949779
Encompass_Preferred
A synthetic nonapeptide sedative; increases EEG delta activity and induces orthodox slow-wave sleep in free-moving rabbits at 30 nmol/kg, i.v.; increases NREM and total sleep in free-moving rats
Catalog No. 50-294-9779 Supplier Cayman Chemical Supplier No. 3442650 MG
A synthetic nonapeptide sedative; increases EEG delta activity and induces orthodox slow-wave sleep in free-moving rabbits at 30 nmol/kg, i.v.; increases NREM and total sleep in free-moving rats