Learn More
Medchemexpress LLC HY-16194 1mg Medchemexpress, Emiglitate CAS:80879-63-6 Purity:>98%
SDP
Catalog No. 502081874
Encompass_Preferred
Medchemexpress, HY-16194 1mg Emiglitate CAS:80879-63-6 Emiglitate (BAY o 1248) is a potent, selective and competitive inhibitor of α-glucoside hydrolase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1874 Supplier Medchemexpress LLC Supplier No. HY161941MG
Medchemexpress, HY-16194 1mg Emiglitate CAS:80879-63-6 Emiglitate (BAY o 1248) is a potent, selective and competitive inhibitor of α-glucoside hydrolase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.