Learn More
Trajan Scientific EMITTER TAPERED TIP, DISTAL COATED 360UM OD X 20UM ID X 50MM, 13 DEG, PK5
SDP
Catalog No. 502725057
Encompass_Preferred
EMITTER TAPERED TIP, DISTAL COATED 360UM OD X 20UM ID X 50MM, 13 DEG, PK5
Catalog No. 50-272-5057 Supplier Trajan Scientific Supplier No. 063240100
EMITTER TAPERED TIP, DISTAL COATED 360UM OD X 20UM ID X 50MM, 13 DEG, PK5