Learn More
Waters Corp WATERS CORPORATION Picotip Pre-cut Emitter, 360μm OD/20μm ID/10μm Tip, 2.5 in. Length
Catalog No. 50465997 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pre-cut Picotip emitter is designed for precision applications requiring low flow rates. It features specific dimensions and a polyimide coating on the outlet tip side.
  • Available in a 5-pack
  • Dimensions: 360μm OD x 20μm ID
  • 10μm tip
  • 2.5 in. length
  • Recommended for flow rates less than 1 μL/min
  • Polyimide coating expected to be removed on the outlet tip side
Catalog No. 50-465-997 Supplier Waters Corp Supplier No. 186003916
This pre-cut Picotip emitter is designed for precision applications requiring low flow rates. It features specific dimensions and a polyimide coating on the outlet tip side.
  • Available in a 5-pack
  • Dimensions: 360μm OD x 20μm ID
  • 10μm tip
  • 2.5 in. length
  • Recommended for flow rates less than 1 μL/min
  • Polyimide coating expected to be removed on the outlet tip side