Learn More
Waters Corp Waters Emitter, Tapertip, Pre-cut, 360μm OD x 20μm ID
Catalog No. 502037666 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pre-cut Tapertip Emitter is designed for precise sample introduction in mass spectrometry applications, particularly suitable for flow rates greater than 1 μL/min. It features a durable polyimide coating and precisely defined dimensions for reliable performance. Supplied in a convenient pack of five, these emitters are essential components for maintaining optimal system operation.
  • Pre-cut for convenient installation.
  • Features a specialized Tapertip design.
  • Constructed with a polyimide coating for durability.
  • Offers precise dimensions of 360μm outer diameter and 20μm inner diameter.
  • Measures 2.5 in. in length.
  • Optimized for flow rates exceeding 1 μL/min.
  • Comes in a pack of 5.
Catalog No. 50-203-7666 Supplier Waters Corp Supplier No. 186003932
This pre-cut Tapertip Emitter is designed for precise sample introduction in mass spectrometry applications, particularly suitable for flow rates greater than 1 μL/min. It features a durable polyimide coating and precisely defined dimensions for reliable performance. Supplied in a convenient pack of five, these emitters are essential components for maintaining optimal system operation.
  • Pre-cut for convenient installation.
  • Features a specialized Tapertip design.
  • Constructed with a polyimide coating for durability.
  • Offers precise dimensions of 360μm outer diameter and 20μm inner diameter.
  • Measures 2.5 in. in length.
  • Optimized for flow rates exceeding 1 μL/min.
  • Comes in a pack of 5.