Learn More
Trajan Scientific EMITTER TAPERED TIP, UNCOATED 360UM OD X 20UM ID X 40MM, 13 DEG, PK5
SDP
Catalog No. 502725061
Encompass_Preferred
EMITTER TAPERED TIP, UNCOATED 360UM OD X 20UM ID X 40MM, 13 DEG, PK5
Catalog No. 50-272-5061 Supplier Trajan Scientific Supplier No. 063200102
EMITTER TAPERED TIP, UNCOATED 360UM OD X 20UM ID X 40MM, 13 DEG, PK5