Learn More
Medchemexpress LLC Emixustat hydrochloride | 1141934-97-5 | 99.29% | 299.84 | 5 MG
SDP
Catalog No. 5000401698
Encompass_Preferred
Emixustat hydrochloride is an orally active RPE65 inhibitor with an IC50 value of 4.4 nM. It is also a visual cycle modulator, capable of regulating visual cycle activity by inhibiting retinol isomerization, and holds potential for studying vision disorders such as age-related macular degeneration (AMD).
  • Orally active RPE65 inhibitor
  • IC50 value of 4.4 nM
  • Visual cycle modulator
  • Inhibits retinol isomerization
  • Regulates visual cycle activity
  • Investigated for potential in studying vision disorders such as age-related macular degeneration (AMD)
Catalog No. 50-004-01698 Supplier Medchemexpress LLC Supplier No. HY19720A5MG
Emixustat hydrochloride is an orally active RPE65 inhibitor with an IC50 value of 4.4 nM. It is also a visual cycle modulator, capable of regulating visual cycle activity by inhibiting retinol isomerization, and holds potential for studying vision disorders such as age-related macular degeneration (AMD).
  • Orally active RPE65 inhibitor
  • IC50 value of 4.4 nM
  • Visual cycle modulator
  • Inhibits retinol isomerization
  • Regulates visual cycle activity
  • Investigated for potential in studying vision disorders such as age-related macular degeneration (AMD)