Learn More
Medchemexpress LLC Emixustat hydrochloride | 1141934-97-5 | 99.3% | 299.84 | 50 MG
SDP
Catalog No. 5000400917
Encompass_Preferred
Emixustat hydrochloride (ACU-4429) is an orally active RPE65 inhibitor with an IC50 value of 4.4 nM. It functions as a visual cycle modulator by inhibiting retinol isomerization. This compound shows potential for research into vision disorders such as age-related macular degeneration (AMD).
  • Reduces the production of 11-cis-retinol in a concentration-dependent manner, thereby inhibiting RPE65 activity.
  • Reduces the conductance of retinal cation channels after dark adaptation in rats.
  • Dose-dependently reduces retinal neovascularization in an Oxygen-Induced Retinopathy (OIR) mouse model.
Catalog No. 50-004-00917 Supplier Medchemexpress LLC Supplier No. HY19720A50MG
Emixustat hydrochloride (ACU-4429) is an orally active RPE65 inhibitor with an IC50 value of 4.4 nM. It functions as a visual cycle modulator by inhibiting retinol isomerization. This compound shows potential for research into vision disorders such as age-related macular degeneration (AMD).
  • Reduces the production of 11-cis-retinol in a concentration-dependent manner, thereby inhibiting RPE65 activity.
  • Reduces the conductance of retinal cation channels after dark adaptation in rats.
  • Dose-dependently reduces retinal neovascularization in an Oxygen-Induced Retinopathy (OIR) mouse model.