Learn More
Medchemexpress LLC Emixustat hydrochloride (ACU-4429 hydrochloride) | 1141934-97-5 | 99.3% | 299.84 | 1 ML
SDP
Catalog No. 5000401256
Encompass_Preferred
Emixustat hydrochloride (ACU-4429 hydrochloride) is an orally active RPE65 inhibitor. It functions as a visual cycle modulator, regulating visual cycle activity by inhibiting retinol isomerization, and shows potential for studying vision disorders, including age-related macular degeneration (AMD).
  • Orally active RPE65 inhibitor
  • Functions as a visual cycle modulator
  • Regulates visual cycle activity by inhibiting retinol isomerization
  • Potential for studying vision disorders such as age-related macular degeneration (AMD)
  • Inhibits RPE65 activity with an IC50 of 4.4 nM
  • Reduces 11-cis-retinol production
  • Shown to inhibit neovascularization and protect the retina in an oxygen-induced retinopathy mouse model
Catalog No. 50-004-01256 Supplier Medchemexpress LLC Supplier No. HY19720A10MM/1ML
Emixustat hydrochloride (ACU-4429 hydrochloride) is an orally active RPE65 inhibitor. It functions as a visual cycle modulator, regulating visual cycle activity by inhibiting retinol isomerization, and shows potential for studying vision disorders, including age-related macular degeneration (AMD).
  • Orally active RPE65 inhibitor
  • Functions as a visual cycle modulator
  • Regulates visual cycle activity by inhibiting retinol isomerization
  • Potential for studying vision disorders such as age-related macular degeneration (AMD)
  • Inhibits RPE65 activity with an IC50 of 4.4 nM
  • Reduces 11-cis-retinol production
  • Shown to inhibit neovascularization and protect the retina in an oxygen-induced retinopathy mouse model