Learn More
ABclonal Technology EML2 Rabbit pAb
SDP
Catalog No. 5000106269
Encompass_Preferred
Enables microtubule binding activity. Involved in negative regulation of microtubule polymerization and regulation of microtubule nucleation. Colocalizes with mitotic spindle.
Catalog No. 50-001-06269 Supplier ABclonal Technology Supplier No. A14218500UL
Enables microtubule binding activity. Involved in negative regulation of microtubule polymerization and regulation of microtubule nucleation. Colocalizes with mitotic spindle.