Learn More
Medchemexpress LLC EML4-ALK kinase inhibitor 1 | 1373409-08-5 | 98.0% | 580.76 | 100 MG
SDP
Catalog No. 5000438894
Encompass_Preferred
EML4-ALK kinase inhibitor 1 is a potent orally active inhibitor of echinoderm microtubule-associated protein-like 4-anaplastic lymphoma kinase (EML4-ALK), with an IC50 of 1 nM.
  • Potent orally active inhibitor of EML4-ALK.
  • IC50 of 1 nM against EML4-ALK.
  • Inhibits EML4-fused ALK variant 1 in mouse BAF3 cells (IC50 1 nM).
  • Exhibits antiproliferative activity against mouse BAF3 cells expressing EML4-fused ALK variant 1 (IC50 9.7 nM).
Catalog No. 50-004-38894 Supplier Medchemexpress LLC Supplier No. HY111752100MG
EML4-ALK kinase inhibitor 1 is a potent orally active inhibitor of echinoderm microtubule-associated protein-like 4-anaplastic lymphoma kinase (EML4-ALK), with an IC50 of 1 nM.
  • Potent orally active inhibitor of EML4-ALK.
  • IC50 of 1 nM against EML4-ALK.
  • Inhibits EML4-fused ALK variant 1 in mouse BAF3 cells (IC50 1 nM).
  • Exhibits antiproliferative activity against mouse BAF3 cells expressing EML4-fused ALK variant 1 (IC50 9.7 nM).