Learn More
Selleck Chemical LLC Emlenoflast-E1213-10MM/1ML
SDP
Catalog No. 503137322
Encompass_Preferred
Emlenoflast a sulfonylurea is a potent and selective inhibitor of NLRP3 inflammasome with an IC50 of 100 nM
Catalog No. 50-313-7322 Supplier Selleck Chemical LLC Supplier No. E121310MM/1ML
Add to Cart
Add to Cart
Emlenoflast a sulfonylurea is a potent and selective inhibitor of NLRP3 inflammasome with an IC50 of 100 nM