Learn More
ACROBiosystems Recombinant Protein;Human EMMPRIN / CD147 Protein, Fc Tag;HEK293;1MG;CD7,Basigin,BSG,5F7,CD147,EMMPRIN,M6,OK,TCSF
Catalog No. 501657946
Encompass_Preferred
Recombinant Protein;Human EMMPRIN / CD147 Protein, Fc Tag;1MG;Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # NP_940991.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.3 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7946 Supplier ACROBiosystems Supplier No. CD7H52591MG
Recombinant Protein;Human EMMPRIN / CD147 Protein, Fc Tag;1MG;Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # NP_940991.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.3 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.