Learn More
ACROBiosystems Recombinant Protein;Human EMMPRIN / CD147 Protein;HEK293;1MG;CD7,Basigin,BSG,5F7,CD147,EMMPRIN,M6,OK,TCSF
Supplier:  ACROBiosystems CD7H52221MG
Encompass_Preferred
Recombinant Protein;Human EMMPRIN / CD147 Protein;1MG;Human EMMPRIN, His Tag (CD7-H5222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # NP_940991.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.6 kDa. The protein migrates as 26-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7511