Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human EMMPRIN / CD147 Protein, Avi Tag (Avitag™);HEK293;200UG;CD7,Basigin,BSG,5F7,CD147,EMMPRIN,M6,OK,TCSF
Catalog No. 501658384
Encompass_Preferred
Recombinant Protein;Biotinylated Human EMMPRIN / CD147 Protein, Avi Tag (Avitag™);200UG;Biotinylated Human EMMPRIN, His Tag (CD7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # P35613-2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 22.8 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8384 Supplier ACROBiosystems Supplier No. CD7H82E0200UG
Recombinant Protein;Biotinylated Human EMMPRIN / CD147 Protein, Avi Tag (Avitag™);200UG;Biotinylated Human EMMPRIN, His Tag (CD7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # P35613-2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 22.8 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.