Learn More
Medchemexpress LLC HY-101476 5mg Medchemexpress, Emodepside CAS:155030-63-0 Purity:>98%
SDP
Catalog No. 501932143
Encompass_Preferred
Medchemexpress, HY-101476 5mg Emodepside CAS:155030-63-0 Emodepside (PF 1022-221) is a cyclooctadepsipeptide with broad-spectrum anthelmintic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2143 Supplier Medchemexpress LLC Supplier No. HY1014765MG
Medchemexpress, HY-101476 5mg Emodepside CAS:155030-63-0 Emodepside (PF 1022-221) is a cyclooctadepsipeptide with broad-spectrum anthelmintic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.