Learn More
Selleck Chemical LLC Emodepside S0467-25MG
SDP
Catalog No. 5000042326
Encompass_Preferred
Emodepside (QHT06 BAY 44-4400 PF 1022-221) a semisynthetic derivative of PF1022A is a cyclooctadepsipeptide with broad-spectrum anthelmintic activity Emodepside activates Ca-dependent SLO-1-like K channels
Catalog No. 50-000-42326 Supplier Selleck Chemical LLC Supplier No. S046725MG
Emodepside (QHT06 BAY 44-4400 PF 1022-221) a semisynthetic derivative of PF1022A is a cyclooctadepsipeptide with broad-spectrum anthelmintic activity Emodepside activates Ca-dependent SLO-1-like K channels