Learn More
Medchemexpress LLC Emodin-1-O-β-D-glucopyranoside | 38840-23-2 | MFCD23379926 | 99.8% | 432.38 g/mol | C21H20O10 | 10 MG
SDP
Catalog No. 5000289342
Encompass_Preferred
Emodin-1-O-β-D-glucopyranoside is a natural anthraquinone glycoside isolated from Polygonum cuspidatum. It acts as a potent, noncompetitive bacterial neuraminidase (BNA) inhibitor (IC50 = 0.85 μM) and is used for biochemical and pharmacological research into neuraminidase activity and natural-product bioactivity.
  • High purity suitable for research (reported purity 99.79%).
  • Well characterized with downloadable data: data sheet, COA, SDS, 1H NMR, LC-MS.
  • Molecular formula C21H20O10 and molecular weight 432.38 g/mol.
  • Available in small research quantities (5 mg, 10 mg, 50 mg, 100 mg).
  • Reported activity supports use in neuraminidase inhibition assays and mechanistic studies.
Catalog No. 50-002-89342 Supplier Medchemexpress LLC Supplier No. HYN239410MG
Emodin-1-O-β-D-glucopyranoside is a natural anthraquinone glycoside isolated from Polygonum cuspidatum. It acts as a potent, noncompetitive bacterial neuraminidase (BNA) inhibitor (IC50 = 0.85 μM) and is used for biochemical and pharmacological research into neuraminidase activity and natural-product bioactivity.
  • High purity suitable for research (reported purity 99.79%).
  • Well characterized with downloadable data: data sheet, COA, SDS, 1H NMR, LC-MS.
  • Molecular formula C21H20O10 and molecular weight 432.38 g/mol.
  • Available in small research quantities (5 mg, 10 mg, 50 mg, 100 mg).
  • Reported activity supports use in neuraminidase inhibition assays and mechanistic studies.