Learn More
AdipoGen Emodin (250 mg)
SDP
Catalog No. 502053628
Encompass_Preferred
Chemical. CAS: 518-82-1. Formula: C15H10O5. MW: 270.2. Isolated from Polygonum cuspidatum. p56lck protein kinase inhibitor. Competitive casein kinase II (CK2) inhibitor. Anticancer agent with apoptosis inducing with antiangiogenic activity. Antioxidant, vasorelaxant and neuroprotective agent. NF-kappaB activation inhibitor. Inflammasome inhibitor. Potent selective 11beta-HSD1 inhibitor. Improves insulin sensitivity and lipid metabolism. Estrogen receptor ERalpha and ERbeta inhibitor. Inhibits androgen receptor (AR) nuclear translocation and induces AR degradation. VEGFR2, FGFR-1, EGFR, PDGFR-beta and Kit inhibitor. JAK2 inhibitor. Anti-diabetic agent. AMPK activator and PPARgamma agonist. Adipogenesis inhibitor. P2X7 receptor antagonist. Wnt signaling & PI3K/Akt/GSK-3beta pathway modulator.
Catalog No. 50-205-3628 Supplier AdipoGen Supplier No. AGCN20457M250
Chemical. CAS: 518-82-1. Formula: C15H10O5. MW: 270.2. Isolated from Polygonum cuspidatum. p56lck protein kinase inhibitor. Competitive casein kinase II (CK2) inhibitor. Anticancer agent with apoptosis inducing with antiangiogenic activity. Antioxidant, vasorelaxant and neuroprotective agent. NF-kappaB activation inhibitor. Inflammasome inhibitor. Potent selective 11beta-HSD1 inhibitor. Improves insulin sensitivity and lipid metabolism. Estrogen receptor ERalpha and ERbeta inhibitor. Inhibits androgen receptor (AR) nuclear translocation and induces AR degradation. VEGFR2, FGFR-1, EGFR, PDGFR-beta and Kit inhibitor. JAK2 inhibitor. Anti-diabetic agent. AMPK activator and PPARgamma agonist. Adipogenesis inhibitor. P2X7 receptor antagonist. Wnt signaling & PI3K/Akt/GSK-3beta pathway modulator.