Learn More
Apexbio Technology LLC Emodin 518-82-1 200mg
SDP
Catalog No. 5000096690
Encompass_Preferred
Emodin (518-82-1) is a small-molecule inhibitor targeting casein kinase 2 (CK2) and the interaction between SARS coronavirus spike protein and angiotensin-converting enzyme 2 (ACE2) It is designed to block protein protein interactions and enzyme activity thereby modulating viral entry and kinase-mediated signaling Emodin exerts its biological activity primarily through inhibition of CK2 and disruption of spike ACE2 binding In hepatocellular carcinoma cell models (Hep3B HepG2 Huh7) emodin demonstrates antiproliferative activity with IC50 values of 66 9 M in Hep3B 74 36 M in HepG2 and 101 5 M in Huh7 cells Based on these pharmacological properties emodin holds research potential in cancer inflammation metabolic disorders and viral infection models
Catalog No. 50-000-96690 Supplier Apexbio Technology LLC Supplier No. A3392200
Emodin (518-82-1) is a small-molecule inhibitor targeting casein kinase 2 (CK2) and the interaction between SARS coronavirus spike protein and angiotensin-converting enzyme 2 (ACE2) It is designed to block protein protein interactions and enzyme activity thereby modulating viral entry and kinase-mediated signaling Emodin exerts its biological activity primarily through inhibition of CK2 and disruption of spike ACE2 binding In hepatocellular carcinoma cell models (Hep3B HepG2 Huh7) emodin demonstrates antiproliferative activity with IC50 values of 66 9 M in Hep3B 74 36 M in HepG2 and 101 5 M in Huh7 cells Based on these pharmacological properties emodin holds research potential in cancer inflammation metabolic disorders and viral infection models