Learn More
Medchemexpress LLC Emodin | 518-82-1 | 99.3% | C15H10O5 | 200 MG
SDP
Catalog No. 5000354292
Encompass_Preferred
Emodin (Frangula emodin) is an anthraquinone derivative known for its anti-SARS-CoV properties. It blocks the SARS coronavirus spike protein and angiotensin-converting enzyme 2 (ACE2) interaction. This compound also inhibits casein kinase-2 (CK2) and demonstrates anti-inflammatory and anticancer effects. Furthermore, it is a potent selective 11β-HSD1 inhibitor and has been shown to ameliorate metabolic disorder in diet-induced obese mice.
  • Blocks SARS coronavirus spike protein and ACE2 interaction
  • Inhibits casein kinase-2 (CK2)
  • Exhibits anti-inflammatory effects
  • Shows anticancer effects
  • Functions as a potent selective 11β-HSD1 inhibitor
  • Ameliorates metabolic disorder in diet-induced obese mice
Catalog No. 50-003-54292 Supplier Medchemexpress LLC Supplier No. HY14393200MG
Emodin (Frangula emodin) is an anthraquinone derivative known for its anti-SARS-CoV properties. It blocks the SARS coronavirus spike protein and angiotensin-converting enzyme 2 (ACE2) interaction. This compound also inhibits casein kinase-2 (CK2) and demonstrates anti-inflammatory and anticancer effects. Furthermore, it is a potent selective 11β-HSD1 inhibitor and has been shown to ameliorate metabolic disorder in diet-induced obese mice.
  • Blocks SARS coronavirus spike protein and ACE2 interaction
  • Inhibits casein kinase-2 (CK2)
  • Exhibits anti-inflammatory effects
  • Shows anticancer effects
  • Functions as a potent selective 11β-HSD1 inhibitor
  • Ameliorates metabolic disorder in diet-induced obese mice