Learn More
Cayman Chemical EmodIn 8-glucosIde 10mg
SDP
Catalog No. 502946965
Encompass_Preferred
An anthraquinone glycoside; inhibits the activities of rat lens aldose reductase and human DNA topoisomerase II (IC50s = 14.4 and 66 μM, respectively, in cell-free assays); promotes the proliferation and differentiation of mouse MC3T3-E1 osteoblastic cells at 1 ng/ml; decreases infarct size in a rat model of focal cerebral ischemia and reperfusion injury at 5 mg/kg
Catalog No. 50-294-6965 Supplier Cayman Chemical Supplier No. 3447410 MG
An anthraquinone glycoside; inhibits the activities of rat lens aldose reductase and human DNA topoisomerase II (IC50s = 14.4 and 66 μM, respectively, in cell-free assays); promotes the proliferation and differentiation of mouse MC3T3-E1 osteoblastic cells at 1 ng/ml; decreases infarct size in a rat model of focal cerebral ischemia and reperfusion injury at 5 mg/kg