Learn More
Medchemexpress LLC Emodin-8-glucoside | 23313-21-5 | MFCD01725365 | 99.4% | 432.38 | C21H20O10 | 10 MG
SDP
Catalog No. 5000296295
Encompass_Preferred
Emodin-8-glucoside is an anthraquinone glycoside (C21H20O10, MW 432.38) supplied as a research reagent. It has reported MAPK inhibitory activity, shows moderate inhibition of rat lens aldose reductase and topoisomerase II, and exhibits antioxidant, anti-inflammatory, and anti-fibrotic properties; it is reported to cross the blood-brain barrier.
  • Anthraquinone glycoside with reported MAPK inhibitory activity.
  • Reported inhibition of aldose reductase and topoisomerase II.
  • Demonstrates antioxidant, anti-inflammatory, and anti-fibrotic effects.
  • High chemical purity suitable for biochemical assays.
  • Intended for laboratory research applications.
Catalog No. 50-002-96295 Supplier Medchemexpress LLC Supplier No. HYN031110MG
Emodin-8-glucoside is an anthraquinone glycoside (C21H20O10, MW 432.38) supplied as a research reagent. It has reported MAPK inhibitory activity, shows moderate inhibition of rat lens aldose reductase and topoisomerase II, and exhibits antioxidant, anti-inflammatory, and anti-fibrotic properties; it is reported to cross the blood-brain barrier.
  • Anthraquinone glycoside with reported MAPK inhibitory activity.
  • Reported inhibition of aldose reductase and topoisomerase II.
  • Demonstrates antioxidant, anti-inflammatory, and anti-fibrotic effects.
  • High chemical purity suitable for biochemical assays.
  • Intended for laboratory research applications.