Learn More
Medchemexpress LLC Emoxypine succinate | 127464-43-1 | 255.27 | 1 G
SDP
Catalog No. 5000449582
Encompass_Preferred
Emoxypine succinate is an antioxidant used for post-traumatic research. It demonstrates reactive oxygen species overproduction and disruption of the mitochondrial inner membrane due to decreasing transmembrane potential. In in vivo studies, it has been shown to decrease the production of reactive oxygen species and the percentage of FITC Annexin V-positive cells of leukocytes.
  • Antioxidant
  • Useful for post-traumatic research
  • Decreases reactive oxygen species production
  • Reduces mitochondrial transmembrane potential
  • Decreases percentage of FITC Annexin V-positive leukocytes
Catalog No. 50-004-49582 Supplier Medchemexpress LLC Supplier No. HYW002620A1G
Emoxypine succinate is an antioxidant used for post-traumatic research. It demonstrates reactive oxygen species overproduction and disruption of the mitochondrial inner membrane due to decreasing transmembrane potential. In in vivo studies, it has been shown to decrease the production of reactive oxygen species and the percentage of FITC Annexin V-positive cells of leukocytes.
  • Antioxidant
  • Useful for post-traumatic research
  • Decreases reactive oxygen species production
  • Reduces mitochondrial transmembrane potential
  • Decreases percentage of FITC Annexin V-positive leukocytes