Learn More
CPC Scientific FITC-LC-Thr-Tyr-Ser-Cys-His-Phe-Gly-Pro-Leu-Thr-Trp-Val-Cys-Lys-Pro-Gln-Gly-Gly-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific SIGN005B
Encompass
This is a fluorescent (FITC)-labeled Erythropoietin (EPO)-mimetic peptide (EMP17), Abs/Em=494/520 nmONE-LETTER SEQUENCE: FITC-LC-TYSCHFGPLTWVCKPQGGMOLECULAR FORMULA: C117H151N25O30S3MOLECULAR WEIGHT:2483.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Hematology
Catalog No. 50-211-0400