Learn More
Aviva Systems Biology Corp EMP2 Antibody - middle region (ARP41470_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41470_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EMP2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human EMP2. Alternate names: MGC9056, XMPApplicationIHC, WB. Species reactivity: Cow, Human
Catalog No. 50-014-53302