Learn More
Medchemexpress LLC HY-108682 1mg Medchemexpress, EMPA CAS:680590-49-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1086821MG
Encompass_Preferred
Medchemexpress, HY-108682 1mg EMPA CAS:680590-49-2 EMPA is a high-affinity, reversible and selective orexin OX2 receptor antagonist. [3H]EMPA binds to human and rat OX2-HEK293 membranes with KD values of 1.1 and 1.4 nM respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8606