Learn More
Medchemexpress LLC HY-15409 10mg , Empagliflozin CAS:864070-44-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1540910MG
Encompass_Preferred
HY-15409 10mg Empagliflozin CAS: 864070-44-0 Empagliflozin (BI 107730 is a selective sodium glucose cotransporter-2 (SGLT-2) inhibitor with an IC50 of 3.1 nM for human SGLT-2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9748