Learn More
AdipoGen Empagliflozin,Chemical. CAS: 864070-44-0
SDP
Supplier:  AdipoGen AGCR13619M010
Encompass_Preferred
Empagliflozin, AdipoGen Life Sciences. Chemical. CAS: 864070-44-0. Formula: C23H27ClO7. MW: 450.9. Antidiabetic agent. Potent and selective sodium glucose co-transporter 2 (SGLT-2) inhibitor. >300-fold selectivity over SGLT-1, 4, 5 and 6. SGLT-2 is found almost exclusively in the proximal tubules of nephronic components in kidneys. Inhibition of SGLT-2 reduces blood glucose by blocking renal glucose reabsorption and thereby increasing urinary glucose excretion (UGE). Shown to preserves beta cell mass and restore glucose homeostasis.
Catalog No. 50-114-6583