Learn More
Medchemexpress LLC HY-15409 100mg , Empagliflozin BI 10773 CAS:864070-44-0 Purity:98%
SDP
Catalog No. 501492717
Encompass_Preferred
Medchemexpress, HY-15409 100mg Empagliflozin BI 10773 CAS:864070-44-0 Formula:C23H27ClO7 IC50: 3.1 nM (SGLT-2), 1.1 μM (SGLT-5), 2 μM (SGLT-6), 8.3 μM (SGLT-1), 11 μM (SGLT-4) Purity:98% Empagliflozin is a selective sodium glucose cotransporter-2 ( SGLT-2 ) inhibitor with an IC 50 of 3.1 nM for hSGLT-2. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2717 Supplier Medchemexpress LLC Supplier No. HY15409100MG
Medchemexpress, HY-15409 100mg Empagliflozin BI 10773 CAS:864070-44-0 Formula:C23H27ClO7 IC50: 3.1 nM (SGLT-2), 1.1 μM (SGLT-5), 2 μM (SGLT-6), 8.3 μM (SGLT-1), 11 μM (SGLT-4) Purity:98% Empagliflozin is a selective sodium glucose cotransporter-2 ( SGLT-2 ) inhibitor with an IC 50 of 3.1 nM for hSGLT-2. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.