Learn More
Selleck Chemical LLC Empagliflozin
SDP
Catalog No. 5000003499
Encompass_Preferred
Empagliflozin is a potent and selective SGLT-2 inhibitor with IC50 of 3 1 nM exhibits 300-fold selectivity over SGLT-1 4 5 and 6 Phase 3
Catalog No. 50-000-03499 Supplier Selleck Chemical LLC Supplier No. S802225MG
Empagliflozin is a potent and selective SGLT-2 inhibitor with IC50 of 3 1 nM exhibits 300-fold selectivity over SGLT-1 4 5 and 6 Phase 3