Learn More
Cayman Chemical Empagliflozin, Purity: ≥98%, CAS: 864070-44-0, 50 mg
SDP
Catalog No. 502967073
Encompass_Preferred
An SGLT2 inhibitor (IC50 = 3.1 nM for the human transporter); reduces non-fasting blood glucose levels, as well as aortic superoxide levels and cardiac interstitial fibrosis, in db/db mice when administered in the diet at 0.03%; decreases the severity of glomerulosclerosis, as well as reduces the escape latency in the Morris water maze, in the same model
Catalog No. 50-296-7073 Supplier Cayman Chemical Supplier No. 1737550 MG
An SGLT2 inhibitor (IC50 = 3.1 nM for the human transporter); reduces non-fasting blood glucose levels, as well as aortic superoxide levels and cardiac interstitial fibrosis, in db/db mice when administered in the diet at 0.03%; decreases the severity of glomerulosclerosis, as well as reduces the escape latency in the Morris water maze, in the same model