Learn More
Apexbio Technology LLC Empagliflozin (BI 10773) 864070-44-0 100mg
SDP
Catalog No. 5000061855
Encompass_Preferred
Empagliflozin (BI 10773 CAS 864070-44-0) is a potent selective inhibitor targeting sodium-glucose cotransporter 2 (SGLT-2) a transmembrane protein primarily expressed in the kidneys and responsible for glucose reabsorption Empagliflozin inhibits SGLT-2 with an IC50 of 3 1 nM demonstrating high selectivity relative to related transporters (SGLT-1 4 5 and 6) In vitro studies using HK2 human proximal tubular cells showed that empagliflozin suppresses high glucose-induced pro-inflammatory signaling pathways including TLR4 NF- B AP-1 and IL-6 secretion Empagliflozin has been widely studied in diabetic animal models such as Zucker diabetic obese rats effectively reducing blood glucose and increasing urinary glucose excretion supporting its application in diabetes research
Catalog No. 50-000-61855 Supplier Apexbio Technology LLC Supplier No. A4601100
Empagliflozin (BI 10773 CAS 864070-44-0) is a potent selective inhibitor targeting sodium-glucose cotransporter 2 (SGLT-2) a transmembrane protein primarily expressed in the kidneys and responsible for glucose reabsorption Empagliflozin inhibits SGLT-2 with an IC50 of 3 1 nM demonstrating high selectivity relative to related transporters (SGLT-1 4 5 and 6) In vitro studies using HK2 human proximal tubular cells showed that empagliflozin suppresses high glucose-induced pro-inflammatory signaling pathways including TLR4 NF- B AP-1 and IL-6 secretion Empagliflozin has been widely studied in diabetic animal models such as Zucker diabetic obese rats effectively reducing blood glucose and increasing urinary glucose excretion supporting its application in diabetes research