Learn More
Medchemexpress LLC Empagliflozin-d4 | 2749293-95-4 | C23H23D4ClO7 | 5 MG
SDP
Catalog No. 5000376651
Encompass_Preferred
Empagliflozin-d4 is deuterium labeled Empagliflozin. Empagliflozin is a selective sodium glucose cotransporter-2 (SGLT-2) inhibitor with an IC50 of 3.1 nM for human SGLT-2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-76651 Supplier Medchemexpress LLC Supplier No. HY15409S5MG
Empagliflozin-d4 is deuterium labeled Empagliflozin. Empagliflozin is a selective sodium glucose cotransporter-2 (SGLT-2) inhibitor with an IC50 of 3.1 nM for human SGLT-2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.