Learn More
Medchemexpress LLC Empagliflozin-d4 | 2749293-95-4 | C23H23D4ClO7 | 500 UG
SDP
Catalog No. 5000376653
Encompass_Preferred
Empagliflozin-d4 is a deuterium-labeled form of Empagliflozin, a selective sodium glucose cotransporter-2 (SGLT-2) inhibitor. This compound is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Selective SGLT-2 inhibitor
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Potential to affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-76653 Supplier Medchemexpress LLC Supplier No. HY15409S500UG
Empagliflozin-d4 is a deuterium-labeled form of Empagliflozin, a selective sodium glucose cotransporter-2 (SGLT-2) inhibitor. This compound is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Selective SGLT-2 inhibitor
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Potential to affect pharmacokinetic and metabolic profiles