Learn More
Cayman Chemical EmpaglIflozIn-d4 500ug
SDP
Catalog No. 502987982
Encompass_Preferred
An internal standard for the quantification of empagliflozin by GC- or LC-MS
Catalog No. 50-298-7982 Supplier Cayman Chemical Supplier No. 22369500 UG
An internal standard for the quantification of empagliflozin by GC- or LC-MS