Learn More
Description
- All-synthetic, multi-mechanism clarifying purifier
- Contains Q-functional anion exchange media
- Includes a fine particle and bioburden reduction membrane
- Helps reduce turbidity, DNA, host cell proteins, and bioburden
- The asymmetric membrane terminates with a 0.2 μm bioburden reduction zone
- Supports monoclonal antibody recovery
- May reduce downstream sterilizing-grade membrane surface area requirements
- May help protect protein A columns by supporting contaminant clearance
- Product components comply with USP Class VI 70°C Biological Reactivity Tests, in vivo
Specifications
Specifications
| Disposable | Single-use |
| Material (Membrane) | Polyamide |
| Pore Size | 0.2 μm |
| Model | BV0.3R |
| Quantity | 4/Cs. |
| Type | Hybrid Purifier |
| Volume (Metric) | 2.0 mL |
| Height (Metric) | 4.8 cm |
| Diameter (Metric) | 4.3 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.