Learn More
MSC Empire Level I-Beam Level, 48 in, 3 Vial, Aluminum
Catalog No. 5000529789
Encompass
This I-beam level is constructed from aluminum with a silver housing. It includes one level vial and two plumb vials, offering a 48-inch length and top-read functionality. Designed for durability, the level features impact-resistant construction and a robust metal frame.
  • Constructed from aluminum
  • 48 in length
  • Equipped with 3 vials
  • Features one level vial and two plumb vials
  • Not magnetic
  • Top-read capability for easy viewing
  • Front graduation location
  • Silver housing color
  • Impact-resistant design
  • Durable metal frame
  • Manufactured in the United States
Catalog No. 50-005-29789 Supplier MSC Supplier No. 02726057
This I-beam level is constructed from aluminum with a silver housing. It includes one level vial and two plumb vials, offering a 48-inch length and top-read functionality. Designed for durability, the level features impact-resistant construction and a robust metal frame.
  • Constructed from aluminum
  • 48 in length
  • Equipped with 3 vials
  • Features one level vial and two plumb vials
  • Not magnetic
  • Top-read capability for easy viewing
  • Front graduation location
  • Silver housing color
  • Impact-resistant design
  • Durable metal frame
  • Manufactured in the United States