Learn More
MSC Empire Level Square, 9 in Blade Length, 6.5 in Base Length, Stainless Steel, Aluminum
Catalog No. 5000528051
Encompass
The Empire Level T-Bevel Square is a versatile tool designed for accurately transferring angle edges for cutting. Constructed with a 9-in stainless steel blade and an aluminum base, this square is built for durability and precision in various workshop and construction applications.
  • Used to transfer angle edges.
  • Features a 9 in blade length.
  • Includes a 6.5 in base length.
  • Constructed with a stainless steel blade for durability.
  • Features a lightweight aluminum base.
  • Ideal for use as an angle finder.
Catalog No. 50-005-28051 Supplier MSC Supplier No. 77589455
The Empire Level T-Bevel Square is a versatile tool designed for accurately transferring angle edges for cutting. Constructed with a 9-in stainless steel blade and an aluminum base, this square is built for durability and precision in various workshop and construction applications.
  • Used to transfer angle edges.
  • Features a 9 in blade length.
  • Includes a 6.5 in base length.
  • Constructed with a stainless steel blade for durability.
  • Features a lightweight aluminum base.
  • Ideal for use as an angle finder.