Learn More
MSC Empire Level Steel Square, 24 inch Blade Length, 16 inch Base Length, 0.06 inch Thick
Catalog No. 5000527312
Encompass
The steel square is a durable marking tool designed for precision in various applications. It features a 24 inch blade and a 16 inch base, constructed from steel with a 0.06 inch thickness, making it suitable for professionals requiring accurate measurements and layouts.
  • Stamped numbers for easy reading
  • Durable protective epoxy coating
  • Designed for use with marking tools
  • 24 inch blade length
  • 16 inch base length
  • 0.06 inch blade thickness
  • 1-1/2 inch base width
  • 2 inch blade width
Catalog No. 50-005-27312 Supplier MSC Supplier No. 64495740
The steel square is a durable marking tool designed for precision in various applications. It features a 24 inch blade and a 16 inch base, constructed from steel with a 0.06 inch thickness, making it suitable for professionals requiring accurate measurements and layouts.
  • Stamped numbers for easy reading
  • Durable protective epoxy coating
  • Designed for use with marking tools
  • 24 inch blade length
  • 16 inch base length
  • 0.06 inch blade thickness
  • 1-1/2 inch base width
  • 2 inch blade width