Learn More
LGC GENOMICS LLC (Lucigen) Ampligase DNA Ligase with Buffer, 750 U @ 5 U/LHighly thermostable DNA ligase for applications where ligation at high temperature is beneficial or required.
Catalog No. NC9779285
Encompass
Ampligase Thermostable DNA Ligase catalyzes NAD-dependent ligation of adjacent 3-hydroxylated and 5-phosphorylated termini in duplex DNA structures that are stable at high temperatures
This item is not returnable. View return policy
Catalog No. NC9779285 Supplier LGC GENOMICS LLC Supplier No. A32750
Add to Cart
Edge
Add to Cart
Ampligase Thermostable DNA Ligase catalyzes NAD-dependent ligation of adjacent 3-hydroxylated and 5-phosphorylated termini in duplex DNA structures that are stable at high temperatures