Learn More
Enzo Life Sciences EMPO (10mg)
SDP
Supplier:  Enzo Life Sciences ALX430098M010
Encompass_Preferred
Beside DEPMPO (ALX-430-093), most efficient spin trap for the in vitro and in vivo detection of O-, N-, S-, and C-centered free radicals. Has a longer life-time than DMPO (ALX-430-090). Alternative name: 2-Ethoxycarbonyl-2-methyl-3,4-dihydro-2H-pyrrole-1-oxide. Purity: ≥95% (HPLC). Solubility: Soluble in water, DMSO, ethanol or dichloromethane. Long Term Storage: -80°C.
Catalog No. 50-200-9201