Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDS Analytical Empore 8000 EZ-Trace SPE Vacuum Manifold
SDP

Catalog No. 01189101 Shop All CDS Analytical Products
Change view
Click to view available options
Voltage:
0/1 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
01-189-101 0/1 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-189-101 Supplier CDS Analytical Supplier No. 98060408017
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Empore EZ-Trace SPE is designed for the improvement of efficiency of traditional disk/cartridge by allowing users to perform four extractions simultaneously, high quality, cost-effective alternative, automated options

  • Up to four extractions performed simultaneously
    • Independent channels design ensures each channel will not effect others
    • Prevents cross-contamination
  • Lure tip fitting at each extraction station make it universally compatible
  • Perfect for traditional SPE cartridges and disk adapter assemblies included
  • Each extraction station equipped with valve for easy switching between sample collection and waste removal.
    • Flow can be stopped with valve to allow for solvent soaking steps
  • Comes with four standard 40 mL collection vials and screw cap adapters for easy setup adn adaptive use of other collection vials
  • Equipped with needle valve for waste and elution flow (precision flow control)
  • For both small and large volume samples
  • Waste switching valve allows for separation of organic and aqueous waste streams
    • Assists in proper disposal of each type of waste
  • Inert sample pathways made of PEEK, PTFE, and stainless steel prevent corrosion from organic solvents
    • Extends application range and lifetime of parts
  • Simple flow design makes it easy to maintain, clean and replace components

Specifications

Description Disk holders approximately 75 mL, but compatible with users sample bottle for large volume sampling, Any size SPE cartridge with standard luer tip fitting
Format Disks and cartridges
For Use With (Application) Small and large volume samples, Environmental, Toxicology, Pharmaceutical, Bioanalytical
For Use With (Equipment) Solid phase extraction consumables
Product Line Empore
Type SPE manifold
Voltage 0/1 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.