Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDS Empore™ Chelating Extraction Disks
SDP

Catalog No. 13110029 Shop All CDS Analytical Products
Change view
Click to view available options
Quantity:
60/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-110-029 60/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-110-029 Supplier CDS Supplier No. 98060402389EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formerly 3M™ Empore™ Chelating Solid Phase Extraction Disk (SPE): 2271. Provide a mechanism to selectively remove multivalent metal cations from solutions

  • pH range 1-14
  • Chelating Resin, PSDB-Iminodiacetic Acid Groups
  • For extraction of Divalent metals and other divalent cations
  • Relative selectivity: Pb>Cu>Cd>Co>Fe>Ca>Sr
  • Capacity is relatively high, dependent on pH and other ions

Specifications

Base Material PS/DVB Resin
For Use With (Application) Extraction of Divalent Metals and Other Divalent Cations
Type Chelating Extraction Disk
Quantity 60/Cs.
Length (Metric) 4.7 cm
Diameter (Metric) Outer 4.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.